1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde 95%
Dodavatel: Apollo Scientific
OR942291-1G
OR942291-250MG
APOSOR942291-1GEA
2690
CZK
APOSOR942291-1G
APOSOR942291-250MG
1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde 95%
1H-Pyrrolo[2,3-c]pyridine-3-carbaldehyde
Vzorec:
C₈H₆N₂O Storage Temperature: Ambient |
MDL Number:
MFCD08272238 CAS číslo: 25957-65-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...