Tisk…

Methyl-(4-methyl-pyridin-2-yl)-amine 95%

Dodavatel: Apollo Scientific

OR942290-1G OR942290-250MG
APOSOR942290-1GEA 24110 CZK
APOSOR942290-1G APOSOR942290-250MG
Methyl-(4-methyl-pyridin-2-yl)-amine 95%
Methyl-(4-methyl-pyridin-2-yl)-amine
Vzorec: C₇H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD00956049
CAS číslo: 45699-12-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR