2-Chloro-3-methylaminopyridine 95%
Dodavatel: Apollo Scientific
OR942076-1G
OR942076-250MG
APOSOR942076-1GEA
20900
CZK
APOSOR942076-1G
APOSOR942076-250MG
2-Chloro-3-methylaminopyridine 95%
2-Chloro-3-methylaminopyridine
Vzorec:
C₆H₇ClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD01646148 CAS číslo: 40932-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...