Tisk…
(3-Amino-4-methoxy-phenyl)-acetonitrile hydrochloride 95%
Dodavatel: Apollo Scientific
OR942036-1G OR942036-250MG
APOSOR942036-1GEA 21490 CZK
APOSOR942036-1G APOSOR942036-250MG
(3-Amino-4-methoxy-phenyl)-acetonitrile hydrochloride 95%
(3-Amino-4-methoxy-phenyl)-acetonitrile hydrochloride
Vzorec: C₉H₁₀N₂O·ClH
MW: 198,649 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26792440
CAS číslo: 857544-25-3