Tisk…

trans-tert-Butyl 3-iodocyclobutylcarbamate

Dodavatel: Apollo Scientific
Varování

OR941827-1G OR941827-250MG OR941827-500MG
APOSOR941827-1GEA 32670 CZK
APOSOR941827-1G APOSOR941827-250MG APOSOR941827-500MG
trans-tert-Butyl 3-iodocyclobutylcarbamate
trans-tert-Butyl 3-iodocyclobutylcarbamate
Vzorec: C₉H₁₆INO₂
MW: 297,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22415232
CAS číslo: 1389264-19-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR