2-(2-Methoxyphenyl)propan-2-ol 95%
Dodavatel: Apollo Scientific
OR941688-1G
OR941688-250MG
APOSOR941688-1GEA
17990
CZK
APOSOR941688-1G
APOSOR941688-250MG
2-(2-Methoxyphenyl)propan-2-ol 95%
2-(2-Methoxyphenyl)propan-2-ol
Vzorec:
C₁₀H₁₄O₂ MW: 166,217 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD12068167 CAS číslo: 21022-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...