1,2,3,4-Tetrahydroisoquinolin-5-ol 95%
Dodavatel: Apollo Scientific
OR941622-1G
OR941622-250MG
APOSOR941622-1GEA
14820
CZK
APOSOR941622-1G
APOSOR941622-250MG
1,2,3,4-Tetrahydroisoquinolin-5-ol 95%
1,2,3,4-Tetrahydroisoquinolin-5-ol
Vzorec:
C₉H₁₁NO MW: 149,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656937 CAS číslo: 102877-50-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...