Tisk…

Methyl 2-bromoacrylate 98%

Dodavatel: Apollo Scientific

OR941547-1G OR941547-5G
APOSOR941547-1GEA 5390 CZK
APOSOR941547-1G APOSOR941547-5G
Methyl 2-bromoacrylate 98%
Methyl 2-bromoacrylate
Vzorec: C₄H₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD08276754
CAS číslo: 4519-46-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR