Tisk…

3'-Ethoxy-4'-methoxyacetophenone 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(3-Ethoxy-4-methoxyphenyl)ethanone

OR941430-1G OR941430-250MG
APOSOR941430-1GEA 1660 CZK
APOSOR941430-1G APOSOR941430-250MG
3'-Ethoxy-4'-methoxyacetophenone 95%
3'-Ethoxy-4'-methoxyacetophenone
Vzorec: C₁₁H₁₄O₃
MW: 194,227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375406
CAS číslo: 31526-71-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR