5-Hydroxyisoindoline-1,3-dione 95%
Dodavatel: Apollo Scientific
OR941420-1G
APOSOR941420-1GEA
1990
CZK
APOSOR941420-1G
5-Hydroxyisoindoline-1,3-dione 95%
5-Hydroxyisoindoline-1,3-dione
Vzorec:
C₈H₅NO₃ MW: 163,13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11101027 CAS číslo: 50727-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...