Tisk…

2-(Methoxymethoxy)benzonitrile 95%

Dodavatel: Apollo Scientific

OR941401-1G OR941401-250MG
APOSOR941401-1GEA 13860 CZK
APOSOR941401-1G APOSOR941401-250MG
2-(Methoxymethoxy)benzonitrile 95%
2-(Methoxymethoxy)benzonitrile
Vzorec: C₉H₉NO₂
MW: 163,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18826226
CAS číslo: 494209-23-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR