1,6-Naphthyridin-4-amine 95%
Dodavatel: Apollo Scientific
OR941384-1G
OR941384-250MG
APOSOR941384-1GEA
24600
CZK
APOSOR941384-1G
APOSOR941384-250MG
1,6-Naphthyridin-4-amine 95%
1,6-Naphthyridin-4-amine
Vzorec:
C₈H₇N₃ MW: 145,161 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01646213 CAS číslo: 28593-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...