2-Phenyl-1-(pyridin-2-yl)ethanone 95%
Dodavatel: Apollo Scientific
OR941362-1G
OR941362-250MG
APOSOR941362-1GEA
22520
CZK
APOSOR941362-1G
APOSOR941362-250MG
2-Phenyl-1-(pyridin-2-yl)ethanone 95%
2-Phenyl-1-(pyridin-2-yl)ethanone
Vzorec:
C₁₃H₁₁NO MW: 197,233 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00023484 CAS číslo: 27049-45-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...