1-Chloro-4-cyclopropoxybenzene 95%
Dodavatel: Apollo Scientific
OR941227-1G
OR941227-250MG
APOSOR941227-1GEA
20900
CZK
APOSOR941227-1G
APOSOR941227-250MG
1-Chloro-4-cyclopropoxybenzene 95%
1-Chloro-4-cyclopropoxybenzene
Vzorec:
C₉H₉ClO Storage Temperature: Ambient |
MDL Number:
MFCD17000492 CAS číslo: 17204-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...