Tisk…

1-(2-Methoxypyridin-4-yl)ethanone 95%

Dodavatel: Apollo Scientific

OR941139-1G OR941139-250MG
APOSOR941139-1GEA 9260 CZK
APOSOR941139-1G APOSOR941139-250MG
1-(2-Methoxypyridin-4-yl)ethanone 95%
1-(2-Methoxypyridin-4-yl)ethanone
Vzorec: C₈H₉NO₂
MW: 151,163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10697715
CAS číslo: 764708-20-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR