Tisk…

Methyl 4-amino-3,5-dimethylbenzoate 95%

Dodavatel: Apollo Scientific

OR941067-5G
APOSOR941067-5GEA 3970 CZK
APOSOR941067-5G
Methyl 4-amino-3,5-dimethylbenzoate 95%
Methyl 4-amino-3,5-dimethylbenzoate
Vzorec: C₁₀H₁₃NO₂
MW: 179,216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833906
CAS číslo: 3095-48-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR