Tisk…

(S)-Piperidin-3-ol 95%

Dodavatel: Apollo Scientific
Varování

OR940988-5G OR940988-25G
APOSOR940988-5GEA 5760 CZK
APOSOR940988-5G APOSOR940988-25G
(S)-Piperidin-3-ol 95%
(S)-Piperidin-3-ol
Vzorec: C₅H₁₁NO
MW: 101,147 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08669819
CAS číslo: 24211-55-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR