Tisk…

2-Cyano-2-cyclopentylideneacetamide 95%

Dodavatel: Apollo Scientific

OR940917-1G OR940917-500MG
APOSOR940917-1GEA 12400 CZK
APOSOR940917-1G APOSOR940917-500MG
2-Cyano-2-cyclopentylideneacetamide 95%
2-Cyano-2-cyclopentylideneacetamide
Vzorec: C₈H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD01923140
CAS číslo: 875-61-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR