Tisk…

4-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%

Dodavatel: Apollo Scientific

OR9409-1G OR9409-5G
APOSOR9409-1GEA 4630 CZK
APOSOR9409-1G APOSOR9409-5G
4-[Ethyl(methyl)carbamoyl]benzeneboronic acid 98%
4-[Ethyl(methyl)carbamoyl]benzeneboronic acid
Vzorec: C₁₀H₁₄BNO₃
MDL Number: MFCD08056368
CAS číslo: 871333-04-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR