Ethyl 2-chloroethanimidoate hydrochloride 95%
Dodavatel: Apollo Scientific
OR940891-1G
OR940891-5G
APOSOR940891-1GEA
9330
CZK
APOSOR940891-1G
APOSOR940891-5G
Ethyl 2-chloroethanimidoate hydrochloride 95%
Ethyl 2-chloroethanimidoate hydrochloride
Vzorec:
C₄H₈ClNO·ClH MW: 158.026 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD10697958 CAS číslo: 36743-66-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...