(5-Methoxy-1H-benzoimidazol-2-yl)-methanol 95%
Dodavatel: Apollo Scientific
Synonyms:
2-(Hydroxymethyl)-5-methoxybenzimidazole
OR940876-1G
OR940876-5G
APOSOR940876-1GEA
1660
CZK
APOSOR940876-1G
APOSOR940876-5G
(5-Methoxy-1H-benzoimidazol-2-yl)-methanol 95%
(5-Methoxy-1H-benzoimidazol-2-yl)-methanol
Vzorec:
C₉H₁₀N₂O₂ MW: 178,188 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01073114 CAS číslo: 20033-99-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...