3-Nitro-1H-indole 95%
Dodavatel: Apollo Scientific
OR940839-1G
OR940839-250MG
APOSOR940839-1GEA
2610
CZK
APOSOR940839-1G
APOSOR940839-250MG
3-Nitro-1H-indole 95%
3-Nitro-1H-indole
Vzorec:
C₈H₆N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD13178651 CAS číslo: 4770-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...