Tisk…

3-Chloro-2,2-dimethylbut-3-enoic acid 95%

Dodavatel: Apollo Scientific

OR940818-1G OR940818-250MG
APOSOR940818-1GEA 6190 CZK
APOSOR940818-1G APOSOR940818-250MG
3-Chloro-2,2-dimethylbut-3-enoic acid 95%
3-Chloro-2,2-dimethylbut-3-enoic acid
Vzorec: C₆H₉ClO₂
MW: 148,587 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093340
CAS číslo: 56663-75-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR