Tisk…

N-Boc-2-(2-hydroxyethyl)morpholine 97%

Dodavatel: Apollo Scientific

OR940722-1G OR940722-250MG OR940722-500MG
APOSOR940722-1GEA 22320 CZK
APOSOR940722-1G APOSOR940722-250MG APOSOR940722-500MG
N-Boc-2-(2-hydroxyethyl)morpholine 97%
N-Boc-2-(2-hydroxyethyl)morpholine
Vzorec: C₁₁H₂₁NO₄
MW: 231,289 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09608014
CAS číslo: 913642-78-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR