3-Oxocyclopentane-1-carbonitrile 95%
Dodavatel: Apollo Scientific
Synonyms:
3-Oxocyclopentanecarbonitrile
OR940018-1G
OR940018-250MG
APOSOR940018-1GEA
11380
CZK
APOSOR940018-1G
APOSOR940018-250MG
3-Oxocyclopentane-1-carbonitrile 95%
3-Oxocyclopentane-1-carbonitrile
Vzorec:
C₆H₇NO MW: 109,126 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21184612 CAS číslo: 41171-91-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...