Tisk…

3-(Carboxymethyl)benzoic acid 95%

Dodavatel: Apollo Scientific

OR939956-1G OR939956-5G
APOSOR939956-1GEA 0 CZK
APOSOR939956-1G APOSOR939956-5G
3-(Carboxymethyl)benzoic acid 95%
3-(Carboxymethyl)benzoic acid
Vzorec: C₉H₈O₄
MW: 180,157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15523567
CAS číslo: 2084-13-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR