Hex-5-yn-1-amine 96%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR939942-250MG
OR939942-1G
APOSOR939942-250MGEA
6350
CZK
APOSOR939942-250MG
APOSOR939942-1G
Hex-5-yn-1-amine 96%
Hex-5-yn-1-amine
Vzorec:
C₆H₁₁N Bod varu: 144 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09923614 CAS číslo: 15252-45-6 UN: 2733 ADR: 3+8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...