4-Chlorobenzo[d]thiazol-2(3H)-one ≥98%
Dodavatel: Apollo Scientific
OR939907-5G
APOSOR939907-5GEA
2400
CZK
APOSOR939907-5G
4-Chlorobenzo[d]thiazol-2(3H)-one ≥98%
4-Chlorobenzo[d]thiazol-2(3H)-one
Vzorec:
C₇H₄ClNOS MW: 185.631 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00796306 CAS číslo: 39205-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...