Tisk…

3-Methoxycyclohexanecarboxylic acid 95%

Dodavatel: Apollo Scientific

OR939882-1G OR939882-5G
APOSOR939882-1GEA 8070 CZK
APOSOR939882-1G APOSOR939882-5G
3-Methoxycyclohexanecarboxylic acid 95%
3-Methoxycyclohexanecarboxylic acid
Vzorec: C₈H₁₄O₃
MW: 158.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00074991
CAS číslo: 99799-10-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR