4-Chloropyridine-2,6-dicarbonitrile 95%
Dodavatel: Apollo Scientific
OR939507-10G
OR939507-1G
OR939507-5G
APOSOR939507-10GEA
19840
CZK
APOSOR939507-10G
APOSOR939507-1G
APOSOR939507-5G
4-Chloropyridine-2,6-dicarbonitrile 95%
4-Chloropyridine-2,6-dicarbonitrile
Vzorec:
C₇H₂ClN₃ MW: 163,564 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00185811 CAS číslo: 55306-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...