2-Mercapto-5-methylpyridine 95%
Dodavatel: Apollo Scientific
Synonyms:
5-Methylpyridine-2(1H)-thione
OR939445-100MG
OR939445-1G
OR939445-250MG
APOSOR939445-100MGEA
2240
CZK
APOSOR939445-100MG
APOSOR939445-1G
APOSOR939445-250MG
2-Mercapto-5-methylpyridine 95%
2-Mercapto-5-methylpyridine
Vzorec:
C₆H₇NS MW: 125.191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02684159 CAS číslo: 18368-58-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...