Tisk…

2-Chloro-5-cyanobenzoic acid 98%

Dodavatel: Apollo Scientific

OR939327-1G OR939327-250MG
APOSOR939327-1GEA 18850 CZK
APOSOR939327-1G APOSOR939327-250MG
2-Chloro-5-cyanobenzoic acid 98%
2-Chloro-5-cyanobenzoic acid
Vzorec: C₈H₄ClNO₂
MW: 181,576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11036379
CAS číslo: 89891-83-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR