4,4-Dimethyl-2-vinyloxazol-5(4h)-one 95%
Dodavatel: Apollo Scientific
OR939274-1G
OR939274-250MG
APOSOR939274-1GEA
19440
CZK
APOSOR939274-1G
APOSOR939274-250MG
4,4-Dimethyl-2-vinyloxazol-5(4h)-one 95%
4,4-Dimethyl-2-vinyloxazol-5(4h)-one
Vzorec:
C₇H₉NO₂ MW: 139,152 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00085288 CAS číslo: 29513-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...