Tisk…

5-Hydroxypentanal 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR939164-1G OR939164-5G
APOSOR939164-1GEA 787 CZK
APOSOR939164-1G APOSOR939164-5G
5-Hydroxypentanal 95%
5-Hydroxypentanal

Cyclic acetal tautomer

Vzorec: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00044411
CAS číslo: 4221-03-8
UN: 1993
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR