2-Ethylbenzyl alcohol 95%
Dodavatel: Apollo Scientific
Synonyms:
(2-Ethylphenyl)methanol
OR938868-10G
OR938868-1G
OR938868-5G
APOSOR938868-10GEA
4300
CZK
APOSOR938868-10G
APOSOR938868-1G
APOSOR938868-5G
2-Ethylbenzyl alcohol 95%
2-Ethylbenzyl alcohol
Vzorec:
C₉H₁₂O MW: 136,191 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093889 CAS číslo: 767-90-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...