Tisk…

N-Formyl-4-(methylamino)benzoic acid 95%

Dodavatel: Apollo Scientific

OR938759-1G OR938759-5G
APOSOR938759-1GEA 9100 CZK
APOSOR938759-1G APOSOR938759-5G
N-Formyl-4-(methylamino)benzoic acid 95%
N-Formyl-4-(methylamino)benzoic acid
Vzorec: C₉H₉NO₃
MW: 179,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00210445
CAS číslo: 51865-84-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR