Tisk…

1,3-Bis(2-chloroethyl)urea 95%

Dodavatel: Apollo Scientific

OR938715-1G OR938715-250MG OR938715-500MG
APOSOR938715-1GEA 1910 CZK
APOSOR938715-1G APOSOR938715-250MG APOSOR938715-500MG
1,3-Bis(2-chloroethyl)urea 95%
1,3-Bis(2-chloroethyl)urea
Vzorec: C₅H₁₀Cl₂N₂O
Storage Temperature: Ambient
MDL Number: MFCD00068573
CAS číslo: 2214-72-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR