6-Methyl-2-vinyl-1,3,6,2-dioxazaborocane-4,8-dione 95%
Dodavatel: Apollo Scientific
OR938711-1G
OR938711-250MG
APOSOR938711-1GEA
4700
CZK
APOSOR938711-1G
APOSOR938711-250MG
6-Methyl-2-vinyl-1,3,6,2-dioxazaborocane-4,8-dione 95%
6-Methyl-2-vinyl-1,3,6,2-dioxazaborocane-4,8-dione
Vzorec:
C₇H₁₀BNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD11656092 CAS číslo: 1104636-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...