Tisk…

7-Methoxyquinazolin-4(1H)-one 95%

Dodavatel: Apollo Scientific

OR938708-1G OR938708-250MG
APOSOR938708-1GEA 3540 CZK
APOSOR938708-1G APOSOR938708-250MG
7-Methoxyquinazolin-4(1H)-one 95%
7-Methoxyquinazolin-4(1H)-one
Vzorec: C₉H₈N₂O₂
MW: 176,172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460971
CAS číslo: 16064-24-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR