Tisk…

Methyl tetrahydrofuran-3-carboxylate 95%

Dodavatel: Apollo Scientific

OR938707-10G OR938707-1G OR938707-5G
APOSOR938707-10GEA 3810 CZK
APOSOR938707-10G APOSOR938707-1G APOSOR938707-5G
Methyl tetrahydrofuran-3-carboxylate 95%
Methyl tetrahydrofuran-3-carboxylate
Vzorec: C₆H₁₀O₃
MW: 130,142 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203928
CAS číslo: 53662-85-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR