1-Methyl-2-oxopyrrolidine-3-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR938615-1G
OR938615-5G
APOSOR938615-1GEA
1080
CZK
APOSOR938615-1G
APOSOR938615-5G
1-Methyl-2-oxopyrrolidine-3-carboxylic acid
1-Methyl-2-oxopyrrolidine-3-carboxylic acid
Vzorec:
C₆H₉NO₃ MW: 143,141 g/mol Bod tání: 92…94 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD19228580 CAS číslo: 30932-84-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...