Tisk…

2-Toluenesulphonyl isocyanate 95%

Dodavatel: Apollo Scientific

OR938567-1G OR938567-25G OR938567-5G
APOSOR938567-1GEA 3540 CZK
APOSOR938567-1G APOSOR938567-25G APOSOR938567-5G
2-Toluenesulphonyl isocyanate 95%
2-Toluenesulphonyl isocyanate

May form small amount of sediment

Vzorec: C₈H₇NO₃S
Storage Temperature: Ambient
MDL Number: MFCD00037045
CAS číslo: 32324-19-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR