(R)-4-Methyloxazolidin-2-one 95%
Dodavatel: Apollo Scientific
OR938550-1G
OR938550-250MG
APOSOR938550-1GEA
1370
CZK
APOSOR938550-1G
APOSOR938550-250MG
(R)-4-Methyloxazolidin-2-one 95%
(R)-4-Methyloxazolidin-2-one
Vzorec:
C₄H₇NO₂ MW: 101.104 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06656588 CAS číslo: 4042-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...