4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Cyano-4-((phenylcarbonothioyl)thio)pentanoic acid
OR938448-1G
OR938448-5G
APOSOR938448-1GEA
1740
CZK
APOSOR938448-1G
APOSOR938448-5G
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid 98%
4-Cyano-4-(phenylcarbonothioylthio)pentanoic acid
Vzorec:
C₁₃H₁₃NO₂S₂ MW: 279,378 g/mol Bod varu: 473 °C Bod tání: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD10698690 CAS číslo: 201611-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...