Tisk…

1-(Azidomethyl)-2-chlorobenzene 95%

Dodavatel: Apollo Scientific

OR938444-1G OR938444-250MG OR938444-5G
APOSOR938444-1GEA 3110 CZK
APOSOR938444-1G APOSOR938444-250MG APOSOR938444-5G
1-(Azidomethyl)-2-chlorobenzene 95%
1-(Azidomethyl)-2-chlorobenzene
Vzorec: C₇H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD11637163
CAS číslo: 63777-70-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR