Tisk…

1,2,3,4-Tetrahydroisoquinolin-8-ol 95%

Dodavatel: Apollo Scientific

OR938375-1G OR938375-250MG
APOSOR938375-1GEA 18190 CZK
APOSOR938375-1G APOSOR938375-250MG
1,2,3,4-Tetrahydroisoquinolin-8-ol 95%
1,2,3,4-Tetrahydroisoquinolin-8-ol
Vzorec: C₉H₁₁NO
MW: 149,19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11506139
CAS číslo: 32999-37-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR