6-Chloro-2,3-dimethylpyridine 95%
Dodavatel: Apollo Scientific
OR938328-1G
OR938328-250MG
APOSOR938328-1GEA
4630
CZK
APOSOR938328-1G
APOSOR938328-250MG
6-Chloro-2,3-dimethylpyridine 95%
6-Chloro-2,3-dimethylpyridine
Vzorec:
C₇H₈ClN MW: 141,598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11505294 CAS číslo: 72093-13-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...