Tisk…

2-Chloro-2,2-dimethyl-2H-1,3-benzoxazine 95%

Dodavatel: Apollo Scientific

OR938278-1G OR938278-250MG OR938278-500MG
APOSOR938278-1GEA 17990 CZK
APOSOR938278-1G APOSOR938278-250MG APOSOR938278-500MG
2-Chloro-2,2-dimethyl-2H-1,3-benzoxazine 95%
2-Chloro-2,2-dimethyl-2H-1,3-benzoxazine
Vzorec: C₁₀H₁₀ClNO
MW: 195,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09840230
CAS číslo: 74405-07-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR