Tisk…

2-Chloro-4-methylbenzothiazole 95%

Dodavatel: Apollo Scientific

OR938249-1G OR938249-250MG
APOSOR938249-1GEA 8930 CZK
APOSOR938249-1G APOSOR938249-250MG
2-Chloro-4-methylbenzothiazole 95%
2-Chloro-4-methylbenzothiazole
Vzorec: C₈H₆ClNS
MW: 183,658 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03840701
CAS číslo: 3622-32-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR