Tisk…

2,5-Dichlorophenylacetyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

OR9377-1G OR9377-5G
APOSOR9377-1GEA 3970 CZK
APOSOR9377-1G APOSOR9377-5G
2,5-Dichlorophenylacetyl chloride
2,5-Dichlorophenylacetyl chloride
Vzorec: C₈H₅Cl₃O
MW: 223,484 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08543437
CAS číslo: 203314-48-1
UN: 3265
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR